Assay Details
Target Gene Details
Entrez Gene ID: | 2120 |
Gene Name: | ETS variant 6 |
Gene Aliases: |
TEL, TEL/ABL, THC5 |
Location: |
Chr.12:11649601-11895402 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ETV6 | NM_001987.4 | 1 | 197 | NP_001978.1 |
| XM_011520607.2 | 1 | 450 | XP_011518909.1 | |
| XM_011520611.2 | 1 | 124 | XP_011518913.1 | |
| XM_017018990.1 | 1 | 450 | XP_016874479.1 | |
| XM_017018991.1 | 1 | 107 | XP_016874480.1 | |
| AK289441.1 | 1 | 65 | ||
| BC043399.1 | 1 | 197 | AAH43399.1 | |
| U11732.1 | AAA19786.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1070094 | Chr.12:11649565 - 11650766 on Build GRCh38 | Deletion |
|
| nsv1046355 | Chr.12:11501836 - 12890185 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map