Assay Details
Target Gene Details
Entrez Gene ID: | 51703 |
Gene Name: | acyl-CoA synthetase long-chain family member 5 |
Gene Aliases: |
ACS2, ACS5, FACL5 |
Location: |
Chr.10:112374158-112428380 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACSL5 | NM_016234.3 | 3 | 491 | NP_057318.2 |
| NM_203379.1 | 3 | 352 | NP_976313.1 | |
| NM_203380.1 | 3 | 518 | NP_976314.1 | |
| XM_017016324.1 | 3 | 526 | XP_016871813.1 | |
| AB033899.1 | 3 | 365 | BAA85979.1 | |
| AK000339.1 | 3 | 501 | ||
| AK024932.1 | 3 | 492 | ||
| AK222782.1 | 3 | 509 | ||
| AK223288.1 | 3 | 480 | ||
| AK310436.1 | 3 | 301 | ||
| AL708480.1 | 3 | 492 | ||
| AM262166.1 | 2 | 211 | ||
| AY358520.1 | 3 | 452 | AAQ88884.1 | |
| BC007985.2 | 3 | 469 | AAH07985.2 | |
| CD689921.1 | 3 | 549 | ||
| CF595490.1 | 3 | 355 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2546876 | Chr.10:112397698 - 112399150 on Build GRCh38 | Deletion |
|
| nsv831990 | Chr.10:112349267 - 112531997 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map