Assay Details
Target Gene Details
Entrez Gene ID: | 23001 |
Gene Name: | WD repeat and FYVE domain containing 3 |
Gene Aliases: |
ALFY, ZFYVE25 |
Location: |
Chr.4:84669537-84966391 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 72 - Exon 72 |
More Information
Set Membership: |
|

Genomic Map