Assay Details
Target Gene Details
Entrez Gene ID: | 284119 |
Gene Name: | polymerase I and transcript release factor |
Gene Aliases: |
CAVIN, CAVIN1, CGL4, FKSG13, cavin-1 |
Location: |
Chr.17:42402449-42423320 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTRF | NM_012232.5 | 1 | 420 | NP_036364.2 |
| XM_005257242.3 | 1 | 420 | XP_005257299.1 | |
| AF312393.1 | 1 | 617 | AAG27093.1 | |
| AK092406.1 | ||||
| AK298072.1 | 1 | 355 | ||
| AK298563.1 | 1 | 356 | ||
| AK314389.1 | 1 | 356 | ||
| BC008849.1 | AAH08849.1 | |||
| BC066123.1 | 1 | 356 | AAH66123.1 | |
| BC073759.1 | 1 | 512 | AAH73759.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470586 | Chr.17:42362183 - 42459270 on Build GRCh38 | Gain |
|
| nsv575051 | Chr.17:42317580 - 42498404 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map