Assay Details
Target Gene Details
Entrez Gene ID: | 10186 |
Gene Name: | lipoma HMGIC fusion partner |
Gene Aliases: |
- |
Location: |
Chr.13:39342892-39603219 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LHFP | NM_005780.2 | 4 | 1170 | NP_005771.1 |
| XM_011534861.1 | 4 | 973 | XP_011533163.1 | |
| AF098807.1 | 4 | 1015 | AAD31386.1 | |
| AK129819.1 | 4 | 671 | ||
| AK223369.1 | 4 | 671 | BAD97089.1 | |
| BC017824.1 | 4 | 1018 | AAH17824.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1036674 | Chr.13:39267345 - 39433980 on Build GRCh38 | Loss |
|
| nsv1054600 | Chr.13:39227146 - 39970890 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map