Assay Details
Target Gene Details
Entrez Gene ID: | 80315 |
Gene Name: | cytoplasmic polyadenylation element binding protein 4 |
Gene Aliases: |
CPE-BP4, hCPEB-4 |
Location: |
Chr.5:173888328-173960991 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPEB4 | NM_001308189.1 | 1 | 1460 | NP_001295118.1 |
| NM_001308191.1 | 1 | 1460 | NP_001295120.1 | |
| NM_030627.3 | 1 | 1460 | NP_085130.2 | |
| XM_005265994.1 | 1 | 1460 | XP_005266051.1 | |
| XM_011534660.2 | 1 | 1460 | XP_011532962.1 | |
| XM_011534661.2 | 1 | 1460 | XP_011532963.1 | |
| AB051460.1 | 1 | 122 | BAB21764.1 | |
| AK310905.1 | 1 | 257 | ||
| BC117150.1 | 1 | 448 | ||
| BC143958.1 | 1 | 448 | ||
| BC143959.1 | 1 | 448 | ||
| BX538213.1 | 1 | 602 | CAD98072.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv600298 | Chr.5:173345250 - 174645532 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map