Assay Details
Target Gene Details
Entrez Gene ID: | 5190 |
Gene Name: | peroxisomal biogenesis factor 6 |
Gene Aliases: |
HMLR2, PAF-2, PAF2, PBD4A, PDB4B, PXAAA1 |
Location: |
Chr.6:42963873-42979243 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PEX6 | NM_000287.3 | NP_000278.3 | ||
| NM_001316313.1 | NP_001303242.1 | |||
| NR_133009.1 | ||||
| XM_011514661.1 | XP_011512963.1 | |||
| AB051076.1 | BAB83046.1 | |||
| AB051077.1 | BAB83047.1 | |||
| AB051078.1 | BAB83048.1 | |||
| AB208906.1 | 12 | 3375 | BAD92143.1 | |
| AK314237.1 | ||||
| BC048331.1 | AAH48331.1 | |||
| D83703.1 | BAA12069.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830651 | Chr.6:42845563 - 43000871 on Build GRCh38 | Loss |
|
| esv3576145 | Chr.6:42728097 - 43012090 on Build GRCh38 | Gain |
|
| nsv602992 | Chr.6:42948182 - 43110718 on Build GRCh38 | Loss |
|
| nsv1017398 | Chr.6:42659705 - 43400998 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map