Assay Details
Target Gene Details
Entrez Gene ID: | 9487 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class L |
Gene Aliases: |
CHIME |
Location: |
Chr.17:16217195-16351800 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGL | NM_004278.3 | NP_004269.1 | ||
| XM_011524080.2 | XP_011522382.1 | |||
| XM_017025349.1 | XP_016880838.1 | |||
| XM_017025350.1 | XP_016880839.1 | |||
| XM_017025351.1 | XP_016880840.1 | |||
| XM_017025352.1 | XP_016880841.1 | |||
| XM_017025353.1 | XP_016880842.1 | |||
| XM_017025354.1 | XP_016880843.1 | |||
| XM_017025355.1 | XP_016880844.1 | |||
| XM_017025356.1 | XP_016880845.1 | |||
| AB017165.1 | BAA74775.1 | |||
| AK292932.1 | ||||
| AK296120.1 | ||||
| AK302523.1 | ||||
| BC068197.1 | AAH68197.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528263 | Chr.17:16194116 - 16293682 on Build GRCh38 | Loss |
|
| nsv522169 | Chr.17:16020395 - 16244507 on Build GRCh38 | Gain |
|
| esv2715702 | Chr.17:15750514 - 16668268 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map