Assay Details
Target Gene Details
Entrez Gene ID: | 11113 |
Gene Name: | citron rho-interacting serine/threonine kinase |
Gene Aliases: |
CRIK, STK21 |
Location: |
Chr.12:119685790-119877318 on Build GRCh38 |
Assay Gene Location: | Within Exon 50 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CIT | NM_001206999.1 | 48 | 6548 | NP_001193928.1 |
| NM_007174.2 | 47 | 6422 | NP_009105.1 | |
| XM_006719206.2 | 48 | 6530 | XP_006719269.1 | |
| XM_011537783.1 | 49 | 6620 | XP_011536085.1 | |
| XM_011537784.1 | 49 | 6617 | XP_011536086.1 | |
| XM_011537785.1 | 49 | 6575 | XP_011536087.1 | |
| XM_011537787.1 | 47 | 6404 | XP_011536089.1 | |
| XM_011537788.1 | 47 | 6401 | XP_011536090.1 | |
| XM_011537789.1 | 41 | 5755 | XP_011536091.1 | |
| XM_011537790.2 | 40 | 5323 | XP_011536092.1 | |
| XM_011537791.2 | 40 | 6308 | XP_011536093.1 | |
| XM_011537792.2 | 39 | 6360 | XP_011536094.1 | |
| XM_017018735.1 | 48 | 6572 | XP_016874224.1 | |
| XM_017018736.1 | 49 | 6572 | XP_016874225.1 | |
| XM_017018737.1 | 48 | 6527 | XP_016874226.1 | |
| XM_017018738.1 | 38 | 5864 | XP_016874227.1 | |
| XM_017018739.1 | 38 | 6636 | XP_016874228.1 | |
| AB023166.2 | 39 | 6360 | BAA76793.2 | |
| AY209000.1 | 13 | 1609 | AAP43922.1 | |
| AY257469.1 | 47 | 6419 | AAP13528.1 | |
| AY681966.1 | 48 | 6548 | AAV87216.1 |
More Information
Set Membership: |
|

Genomic Map