Assay Details
Target Gene Details
Entrez Gene ID: | 27179 |
Gene Name: | interleukin 36, alpha |
Gene Aliases: |
FIL1, FIL1(EPSILON), FIL1E, IL-1F6, IL1(EPSILON), IL1F6 |
Location: |
Chr.2:113005461-113011071 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL36A | NM_014440.2 | NP_055255.1 | ||
| XM_005263639.2 | XP_005263696.1 | |||
| XM_011510965.1 | XP_011509267.1 | |||
| XM_017003806.1 | XP_016859295.1 | |||
| AF201831.1 | AAF25211.1 | |||
| AK314149.1 | ||||
| BC107042.1 | AAI07043.1 | |||
| BC107043.1 | AAI07044.1 |
More Information
Set Membership: |
|

Genomic Map