Assay Details
Target Gene Details
Entrez Gene ID: | 27022 |
Gene Name: | forkhead box D3 |
Gene Aliases: |
AIS1, Genesis, HFH2, VAMAS2 |
Location: |
Chr.1:63323059-63325126 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXD3 | NM_012183.2 | 1 | 1138 | NP_036315.1 |
Target Gene Details
Entrez Gene ID: | 100996301 |
Gene Name: | FOXD3 antisense RNA 1 (head to head) |
Gene Aliases: |
pasFOXD3 |
Location: |
Chr.1:63320884-63324441 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXD3-AS1 | NR_121637.1 | |||
| KF925349.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470714 | Chr.1:63301926 - 63537185 on Build GRCh38 | Gain |
|
| nsv546433 | Chr.1:63288065 - 63537185 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map