Assay Details
Target Gene Details
Entrez Gene ID: | 100528018 |
Gene Name: | ARL2-SNX15 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.11:65014113-65040572 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARL2-SNX15 | NR_037650.1 | 10 | 1409 | |
| AK096447.1 | 10 | 1343 |
Target Gene Details
Entrez Gene ID: | 29907 |
Gene Name: | sorting nexin 15 |
Gene Aliases: |
HSAF001435 |
Location: |
Chr.11:65027408-65040572 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNX15 | NM_013306.4 | 7 | 885 | NP_037438.2 |
| NM_147777.3 | NP_680086.2 | |||
| AB209439.1 | 6 | 3661 | BAD92676.1 | |
| AF001435.1 | 7 | 814 | AAB81205.1 | |
| AF175267.2 | 7 | 794 | AAF89955.1 | |
| AF175268.1 | AAF89956.1 | |||
| AK315034.1 | 7 | 816 | ||
| AK315642.1 | 7 | 826 | ||
| BC009897.2 | 7 | 803 | AAH09897.1 | |
| BC012767.2 | 7 | 855 | AAH12767.1 | |
| BC014520.2 | 7 | 855 | AAH14520.1 | |
| BE547440.1 | ||||
| BE884806.1 | ||||
| BT006631.1 | 7 | 755 | AAP35277.1 | |
| BX444780.2 | 2 | 1042 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526181 | Chr.11:64870569 - 65489858 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map