Assay Details
Target Gene Details
Entrez Gene ID: | 3615 |
Gene Name: | inosine monophosphate dehydrogenase 2 |
Gene Aliases: |
IMPD2, IMPDH-II |
Location: |
Chr.3:49024325-49029750 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IMPDH2 | NM_000884.2 | 5 | 503 | NP_000875.2 |
| XM_006713128.3 | 5 | 811 | XP_006713191.1 | |
| XM_017006349.1 | 4 | 688 | XP_016861838.1 | |
| XM_017006350.1 | 4 | 688 | XP_016861839.1 | |
| AY545560.1 | 2 | 121 | AAS50155.1 | |
| BC006124.1 | 5 | 451 | AAH06124.1 | |
| BC012840.1 | 5 | 444 | AAH12840.1 | |
| BC015567.2 | 5 | 442 | AAH15567.1 | |
| J04208.1 | 5 | 458 | AAA36112.1 |
More Information
Set Membership: |
|

Genomic Map