Assay Details
Target Gene Details
Entrez Gene ID: | 84106 |
Gene Name: | PML-RARA regulated adaptor molecule 1 |
Gene Aliases: |
PML-RAR, PRAM-1 |
Location: |
Chr.19:8490055-8502654 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRAM1 | NM_032152.4 | 2 | 179 | NP_115528.4 |
| XM_005272502.2 | 2 | 165 | XP_005272559.1 | |
| XM_011528352.2 | 2 | 185 | XP_011526654.1 | |
| XM_011528353.2 | 2 | 183 | XP_011526655.1 | |
| AJ272324.1 | CAC17767.1 | |||
| AK308227.1 | 2 | 162 | ||
| BC028012.1 | 3 | 136 | AAH28012.1 | |
| BI907455.1 | 2 | 144 | ||
| DA350460.1 | 2 | 165 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv137n111 | Chr.19:8488438 - 8590905 on Build GRCh38 | Deletion |
|
| dgv6258n54 | Chr.19:8487389 - 8564616 on Build GRCh38 | Loss |
|
| nsv578559 | Chr.19:8483291 - 8532981 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78618855] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map