Assay Details
Target Gene Details
Entrez Gene ID: | 219931 |
Gene Name: | two pore segment channel 2 |
Gene Aliases: |
SHEP10, TPC2 |
Location: |
Chr.11:69048876-69090604 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv468620 | Chr.11:69037338 - 69088127 on Build GRCh38 | Loss |
|
| nsv555301 | Chr.11:69053408 - 69093190 on Build GRCh38 | Loss |
|
| nsv469967 | Chr.11:69036983 - 69132346 on Build GRCh38 | Loss |
|
| nsv832196 | Chr.11:68995114 - 69165820 on Build GRCh38 | Loss |
|
| nsv825970 | Chr.11:69015999 - 69088972 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map