Assay Details
Target Gene Details
Entrez Gene ID: | 54941 |
Gene Name: | ring finger protein 125 |
Gene Aliases: |
TNORS, TRAC-1, TRAC1 |
Location: |
Chr.18:32018203-32090751 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF125 | NM_017831.3 | 1 | 322 | NP_060301.2 |
| XM_011526045.2 | 1 | 591 | XP_011524347.1 | |
| XM_011526046.2 | XP_011524348.1 | |||
| XM_011526047.2 | 1 | 601 | XP_011524349.1 | |
| AK000463.1 | 1 | 322 | BAA91182.1 | |
| AK225518.1 | 1 | 322 | ||
| BI666910.1 | 1 | 437 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524199 | Chr.18:32015201 - 32018846 on Build GRCh38 | Loss |
|
| nsv1065285 | Chr.18:31947921 - 32035023 on Build GRCh38 | Loss |
|
| esv3893087 | Chr.18:31984928 - 32067906 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map