Assay Details
Target Gene Details
Entrez Gene ID: | 11021 |
Gene Name: | RAB35, member RAS oncogene family |
Gene Aliases: |
H-ray, RAB1C, RAY |
Location: |
Chr.12:120095094-120116839 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB35 | NM_001167606.1 | 5 | 1004 | NP_001161078.1 |
| NM_006861.6 | 6 | 1129 | NP_006852.1 | |
| XM_011537779.1 | 6 | 1066 | XP_011536081.1 | |
| XM_017018723.1 | 5 | 906 | XP_016874212.1 | |
| AK125543.1 | 7 | 1904 | ||
| AK298391.1 | 2 | 524 | ||
| AK300127.1 | 4 | 653 | ||
| AK304620.1 | ||||
| BC015931.2 | 6 | 1044 | AAH15931.1 | |
| BP303346.1 | 1 | 255 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952492 | Chr.12:120090097 - 120106496 on Build GRCh38 | Deletion |
|
| nsv560433 | Chr.12:120095892 - 120238287 on Build GRCh38 | Loss |
|
| nsv560434 | Chr.12:120096343 - 120107914 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map