Assay Details
Target Gene Details
Entrez Gene ID: | 1493 |
Gene Name: | cytotoxic T-lymphocyte associated protein 4 |
Gene Aliases: |
ALPS5, CD, CD152, CELIAC3, CTLA-4, GRD4, GSE, IDDM12 |
Location: |
Chr.2:203867788-203873960 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTLA4 | NM_001037631.2 | NP_001032720.1 | ||
| NM_005214.4 | NP_005205.2 | |||
| AF414120.1 | AAL07473.1 | |||
| AF486806.1 | AAL96664.1 | |||
| AK313732.1 | ||||
| AY209009.1 | AAO17066.1 | |||
| AY792514.1 | AAV66331.1 | |||
| AY999702.1 | AAY00166.1 | |||
| BC069566.1 | AAH69566.1 | |||
| BC070162.1 | AAH70162.1 | |||
| BC074842.2 | AAH74842.1 | |||
| BC074893.2 | AAH74893.1 | |||
| CD639535.1 | ||||
| DQ785106.1 | ||||
| L15006.1 | AAB59385.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001799 | Chr.2:202908732 - 205615330 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map