Assay Details
Target Gene Details
Entrez Gene ID: | 83873 |
Gene Name: | G protein-coupled receptor 61 |
Gene Aliases: |
BALGR, GPCR3 |
Location: |
Chr.1:109537539-109547260 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR61 | NM_031936.4 | 2 | 1968 | NP_114142.3 |
| XM_017002438.1 | 2 | 4301 | XP_016857927.1 | |
| AF258342.1 | 3 | 1604 | AAK97826.1 | |
| AF317652.1 | AAK12637.1 | |||
| AK290027.1 | 2 | 1968 | ||
| BC067464.1 | 1 | 1354 | AAH67464.1 | |
| JF810889.1 | 1 | 1285 |
More Information
Set Membership: |
|

Genomic Map