Assay Details
Target Gene Details
Entrez Gene ID: | 3281 |
Gene Name: | heat shock factor binding protein 1 |
Gene Aliases: |
NPC-A-13 |
Location: |
Chr.16:83807903-83813002 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSBP1 | NM_001537.3 | 4 | 637 | NP_001528.1 |
| AF068754.1 | AAC25186.1 | |||
| AK172732.1 | 4 | 502 | ||
| AY320409.1 | AAP73809.1 | |||
| BC007515.2 | AAH07515.1 | |||
| BX366956.2 | 4 | 569 | ||
| BX537440.1 | 4 | 524 | CAD97682.1 | |
| BX648972.1 | 4 | 521 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv573401 | Chr.16:83679394 - 83940461 on Build GRCh38 | Gain |
|
| nsv1907 | Chr.16:83800792 - 83835506 on Build GRCh38 | Insertion |
|
| dgv3054n100 | Chr.16:83519838 - 83940461 on Build GRCh38 | Gain |
|
| nsv573398 | Chr.16:83516215 - 83940461 on Build GRCh38 | Gain |
|
| nsv1064977 | Chr.16:83476129 - 83856075 on Build GRCh38 | Loss |
|
| nsv573405 | Chr.16:83789794 - 83850383 on Build GRCh38 | Gain |
|
| nsv833308 | Chr.16:83742201 - 83897750 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map