Assay Details
Target Gene Details
Entrez Gene ID: | 353116 |
Gene Name: | Rab interacting lysosomal protein like 1 |
Gene Aliases: |
GOSPEL, RLP1 |
Location: |
Chr.12:123471073-123533718 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RILPL1 | NM_001319243.1 | 8 | 1942 | NP_001306172.1 |
| NM_001319244.1 | 7 | 1403 | NP_001306173.1 | |
| NM_001319302.1 | 8 | 2353 | NP_001306231.1 | |
| NM_178314.4 | 7 | 1854 | NP_847884.2 | |
| AK096697.1 | 9 | 1784 | BAC04845.1 | |
| BC080626.1 | 7 | 1403 | AAH80626.1 | |
| BC089444.1 | 7 | 1656 | AAH89444.1 |
Target Gene Details
Entrez Gene ID: | 11066 |
Gene Name: | small nuclear ribonucleoprotein U11/U12 subunit 35 |
Gene Aliases: |
HM-1, U1SNRNPBP |
Location: |
Chr.12:123457644-123472362 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNRNP35 | NR_104103.1 | 2 | 1154 |
More Information
Set Membership: |
|

Genomic Map