Assay Details
Target Gene Details
Entrez Gene ID: | 26224 |
Gene Name: | F-box and leucine rich repeat protein 3 |
Gene Aliases: |
FBL3, FBL3A, FBXL3A |
Location: |
Chr.13:76992597-77027196 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXL3 | NM_012158.2 | 5 | 1434 | NP_036290.1 |
| XM_005266336.1 | 5 | 1217 | XP_005266393.1 | |
| XM_005266337.2 | 4 | 1069 | XP_005266394.1 | |
| XM_017020538.1 | XP_016876027.1 | |||
| AF126028.1 | 4 | 1038 | AAF37383.1 | |
| AF129532.1 | 5 | 1101 | AAF04466.1 | |
| AK025484.1 | 3 | 632 | ||
| AK314442.1 | 5 | 1402 | ||
| AL833187.1 | 5 | 1382 | ||
| BC072448.1 | 5 | 1388 | AAH72448.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832653 | Chr.13:76948899 - 77115221 on Build GRCh38 | Loss |
|
| nsv832652 | Chr.13:76855107 - 77041507 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map