Assay Details
Target Gene Details
Entrez Gene ID: | 3700 |
Gene Name: | inter-alpha-trypsin inhibitor heavy chain family member 4 |
Gene Aliases: |
GP120, H4P, IHRP, ITI-HC4, ITIHL1, PK-120, PK120 |
Location: |
Chr.3:52812990-52830701 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITIH4 | NM_001166449.1 | 9 | 1169 | NP_001159921.1 |
| NM_002218.4 | 9 | 1169 | NP_002209.2 | |
| AB209388.1 | 8 | 2301 | BAD92625.1 | |
| AK298407.1 | 8 | 1104 | ||
| AK298412.1 | 9 | 1140 | ||
| AK298429.1 | 9 | 1140 | ||
| AK303769.1 | 8 | 995 | ||
| BC136392.1 | 9 | 1141 | ||
| BC136393.1 | 9 | 1141 | ||
| BX647322.1 | 6 | 682 | ||
| D38595.1 | 9 | 1143 | BAA07602.1 |
Target Gene Details
Entrez Gene ID: | 100873993 |
Gene Name: | ITIH4 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:52823935-52825314 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITIH4-AS1 | NR_046615.1 | 2 | 199 | |
| AI004860.1 | 2 | 199 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv590308 | Chr.3:52820319 - 52826790 on Build GRCh38 | Loss |
|
| nsv508928 | Chr.3:52769883 - 52888575 on Build GRCh38 | Insertion |
|
| esv2762328 | Chr.3:52740868 - 52968134 on Build GRCh38 | Loss |
|
| nsv834698 | Chr.3:52715967 - 52894203 on Build GRCh38 | Gain |
|
| nsv3822 | Chr.3:52799815 - 52832115 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map