Assay Details
Target Gene Details
Entrez Gene ID: | 81555 |
Gene Name: | Yip1 domain family member 5 |
Gene Aliases: |
FinGER5, SB140, SMAP-5, SMAP5, YIP1A |
Location: |
Chr.5:144158159-144170714 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YIPF5 | NM_001024947.3 | 6 | 1153 | NP_001020118.1 |
| NM_001271732.1 | 5 | 1033 | NP_001258661.1 | |
| NM_030799.8 | 6 | 902 | NP_110426.4 | |
| AB014733.1 | 2 | 173 | BAB20270.1 | |
| AF140225.1 | 5 | 712 | AAG48521.1 | |
| AF318329.1 | 6 | 791 | AAL55836.1 | |
| AK054576.1 | 6 | 839 | BAB70763.1 | |
| AK124065.1 | 6 | 1060 | ||
| AK223515.1 | 6 | 765 | BAD97235.1 | |
| AK307251.1 | 6 | 521 | ||
| AY037152.1 | 6 | 834 | AAK67644.1 | |
| AY358863.1 | 6 | 786 | AAQ89222.1 | |
| AY640925.1 | 6 | 818 | AAV51256.1 | |
| AY640926.1 | 6 | 818 | AAV51257.1 | |
| AY640927.1 | 6 | 818 | AAV51258.1 | |
| AY640928.1 | 6 | 1069 | AAV51259.1 | |
| AY640929.1 | 5 | 949 | ||
| AY640934.1 | AAV51260.1 | |||
| BC007829.2 | 6 | 820 | AAH07829.1 | |
| BC014253.2 | 6 | 840 | AAH14253.1 | |
| BC024737.1 | 6 | 837 | AAH24737.1 | |
| BX647859.1 | 6 | 842 | ||
| CR749463.1 | 6 | 841 | CAH18295.1 |
More Information
Set Membership: |
|

Genomic Map