Assay Details
Target Gene Details
Entrez Gene ID: | 27091 |
Gene Name: | calcium voltage-gated channel auxiliary subunit gamma 5 |
Gene Aliases: |
- |
Location: |
Chr.17:66877273-66885278 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNG5 | NM_145811.2 | NP_665810.1 | ||
| AF361351.1 | AAL50046.1 | |||
| AF458898.1 | AAM00595.1 | |||
| AK290171.1 | ||||
| BC101607.1 | AAI01608.1 | |||
| BC101633.1 | AAI01634.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833521 | Chr.17:66858900 - 67027619 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map