Assay Details
Target Gene Details
Entrez Gene ID: | 199786 |
Gene Name: | family with sequence similarity 129 member C |
Gene Aliases: |
BCNP1 |
Location: |
Chr.19:17523301-17556755 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422429 | Chr.19:17529868 - 17579053 on Build GRCh38 | Duplication |
|

Genomic Map