Assay Details
Target Gene Details
Entrez Gene ID: | 25788 |
Gene Name: | RAD54 homolog B (S. cerevisiae) |
Gene Aliases: |
RDH54 |
Location: |
Chr.8:94371960-94475115 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD54B | NM_001205263.1 | 10 | 2359 | NP_001192192.1 |
| NM_012415.3 | 12 | 2336 | NP_036547.1 | |
| AF112481.1 | 12 | 2285 | AAD34331.1 | |
| AK290437.1 | 12 | 2306 | ||
| AK307516.1 | 10 | 2032 | ||
| AL133578.1 | 6 | 1217 | CAB63724.1 | |
| BC001965.2 | 12 | 2278 | AAH01965.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1031480 | Chr.8:94363468 - 94435179 on Build GRCh38 | Gain |
|
| nsv831401 | Chr.8:94206069 - 94381740 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map