Assay Details
Target Gene Details
Entrez Gene ID: | 5571 |
Gene Name: | protein kinase AMP-activated non-catalytic subunit gamma 1 |
Gene Aliases: |
AMPKG |
Location: |
Chr.12:49002272-49018975 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKAG1 | NM_001206709.1 | 12 | 1559 | NP_001193638.1 |
| NM_001206710.1 | 12 | 1642 | NP_001193639.1 | |
| NM_002733.4 | 12 | 1532 | NP_002724.1 | |
| XM_005269019.4 | 11 | 1551 | XP_005269076.1 | |
| XM_005269020.2 | 12 | 1508 | XP_005269077.1 | |
| XM_006719499.2 | 12 | 1650 | XP_006719562.1 | |
| XM_011538562.2 | 13 | 1672 | XP_011536864.1 | |
| XM_017019622.1 | 12 | 1694 | XP_016875111.1 | |
| XM_017019623.1 | 13 | 1718 | XP_016875112.1 | |
| AK097606.1 | 12 | 1492 | BAC05117.1 | |
| AK307994.1 | 12 | 1455 | ||
| BC000358.2 | 12 | 1456 | AAH00358.1 | |
| BM667691.1 | ||||
| BM972308.1 | 1 | 213 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832404 | Chr.12:48864777 - 49064772 on Build GRCh38 | Gain |
|
| nsv558833 | Chr.12:48965097 - 49085029 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map