Assay Details
Target Gene Details
Entrez Gene ID: | 3619 |
Gene Name: | inner centromere protein |
Gene Aliases: |
- |
Location: |
Chr.11:62123973-62155474 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INCENP | NM_001040694.1 | 4 | 1064 | NP_001035784.1 |
| NM_020238.2 | 4 | 1064 | NP_064623.2 | |
| XM_006718533.2 | 4 | 1026 | XP_006718596.1 | |
| XM_011544995.2 | 4 | 1026 | XP_011543297.1 | |
| XM_011544996.2 | 4 | 1026 | XP_011543298.1 | |
| XM_011544997.2 | 4 | 986 | XP_011543299.1 | |
| XM_011544998.2 | 4 | 1026 | XP_011543300.1 | |
| AF282265.1 | 3 | 862 | AAF87584.1 | |
| AK056195.1 | 4 | 1039 | ||
| AK307633.1 | 4 | 1081 | ||
| AK310714.1 | 4 | 1026 | ||
| AY714053.1 | 3 | 862 | AAU04398.1 | |
| BC098576.1 | 4 | 1064 | AAH98576.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952460 | Chr.11:62127329 - 62154428 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map