Assay Details
Target Gene Details
Entrez Gene ID: | 6039 |
Gene Name: | ribonuclease A family member k6 |
Gene Aliases: |
RAD1, RNS6, RNasek6 |
Location: |
Chr.14:20780956-20782467 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNASE6 | NM_005615.4 | 2 | 379 | NP_005606.1 |
| XM_017021566.1 | 3 | 266 | XP_016877055.1 | |
| XM_017021567.1 | 3 | 279 | XP_016877056.1 | |
| AK313580.1 | 2 | 362 | ||
| BC020848.1 | 2 | 379 | AAH20848.1 | |
| CR456965.1 | 1 | 86 | CAG33246.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759972 | Chr.14:20747949 - 21003455 on Build GRCh38 | Gain+Loss |
|
| esv23438 | Chr.14:20332606 - 20789719 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map