Assay Details
Target Gene Details
Entrez Gene ID: | 5934 |
Gene Name: | RB transcriptional corepressor like 2 |
Gene Aliases: |
P130, Rb2 |
Location: |
Chr.16:53434420-53491648 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBL2 | NM_001323608.1 | 2 | 435 | NP_001310537.1 |
| NM_001323609.1 | 2 | 435 | NP_001310538.1 | |
| NM_001323610.1 | 2 | 435 | NP_001310539.1 | |
| NM_001323611.1 | 2 | 184 | NP_001310540.1 | |
| NM_005611.3 | 2 | 416 | NP_005602.3 | |
| AK300047.1 | 2 | 154 | ||
| AK307881.1 | 2 | 393 | ||
| BC034490.1 | 2 | 406 | AAH34490.1 | |
| DB017942.1 | 2 | 416 | ||
| S67171.1 | 2 | 326 | AAB29227.1 | |
| X74594.1 | 2 | 127 | CAA52671.1 | |
| X76061.1 | 2 | 367 | CAA53661.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3638649 | Chr.16:53427199 - 53440126 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map