Assay Details
Target Gene Details
Entrez Gene ID: | 390261 |
Gene Name: | olfactory receptor family 6 subfamily M member 1 |
Gene Aliases: |
OR11-271 |
Location: |
Chr.11:123805408-123806349 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR6M1 | NM_001005325.1 | 1 | 851 | NP_001005325.1 |
| BC136929.1 | 1 | 889 | ||
| BC136931.1 | 1 | 889 |
More Information
Set Membership: |
|

Genomic Map