Assay Details
Target Gene Details
Entrez Gene ID: | 431705 |
Gene Name: | astacin-like metallo-endopeptidase (M12 family) |
Gene Aliases: |
SAS1B |
Location: |
Chr.2:96122817-96143059 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASTL | NM_001002036.3 | 9 | 1178 | NP_001002036.3 |
| XM_011511205.2 | 8 | 1993 | XP_011509507.1 | |
| XM_011511206.2 | 9 | 2216 | XP_011509508.1 | |
| XM_011511207.2 | 8 | 1940 | XP_011509509.1 | |
| AJ537600.2 | 9 | 1178 | CAD61265.2 | |
| BC107126.1 | 9 | 1177 | AAI07127.1 | |
| BC107127.1 | 9 | 1178 | AAI07128.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv607e201 | Chr.2:95855884 - 97561622 on Build GRCh38 | Deletion |
|
| dgv4009n100 | Chr.2:96072837 - 96159625 on Build GRCh38 | Gain |
|
| nsv1135873 | Chr.2:95981454 - 97579645 on Build GRCh38 | Deletion |
|
| esv3425717 | Chr.2:95859900 - 97515905 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map