Assay Details
Target Gene Details
Entrez Gene ID: | 58538 |
Gene Name: | membrane palmitoylated protein 4 |
Gene Aliases: |
ALS2CR5, DLG6 |
Location: |
Chr.2:201644874-201700263 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPP4 | NM_033066.2 | 22 | 2070 | NP_149055.1 |
| XM_017004620.1 | 18 | 3546 | XP_016860109.1 | |
| AB053302.1 | 18 | 1938 | BAB69012.1 | |
| AB053304.1 | 13 | 2140 | ||
| AF316032.1 | 22 | 2070 | AAK71862.1 | |
| AK131208.1 | 19 | 1983 | BAD18399.1 | |
| AK298589.1 | 17 | 1925 | ||
| AK298947.1 | 19 | 1955 | ||
| AK300622.1 | 17 | 1863 | ||
| BC132785.1 | 20 | 1946 | ||
| BC144206.1 | 17 | 1874 | ||
| BM999928.1 | 1 | 406 | ||
| BX647162.1 | 25 | 4535 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3584274 | Chr.2:201624984 - 201666019 on Build GRCh38 | Loss |
|
| dgv7249n54 | Chr.2:201644883 - 201666824 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map