Assay Details
Target Gene Details
Entrez Gene ID: | 23122 |
Gene Name: | cytoplasmic linker associated protein 2 |
Gene Aliases: |
- |
Location: |
Chr.3:33496245-33718356 on Build GRCh38 |
Assay Gene Location: | Within Exon 39 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv460487 | Chr.3:33468269 - 33561415 on Build GRCh38 | Gain |
|
| nsv1001132 | Chr.3:33386293 - 33565948 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map