Assay Details
Target Gene Details
Entrez Gene ID: | 832 |
Gene Name: | capping actin protein of muscle Z-line beta subunit |
Gene Aliases: |
CAPB, CAPPB, CAPZ |
Location: |
Chr.1:19338773-19485641 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAPZB | NM_001206540.2 | 5 | 635 | NP_001193469.1 |
| NM_001206541.2 | 6 | 1132 | NP_001193470.1 | |
| NM_001282162.1 | 5 | 947 | NP_001269091.1 | |
| NM_004930.4 | 5 | 635 | NP_004921.1 | |
| XM_006710938.3 | 5 | 528 | XP_006711001.1 | |
| XM_011542228.2 | 5 | 1456 | XP_011540530.1 | |
| XM_011542229.2 | 4 | 643 | XP_011540531.1 | |
| XM_011542230.2 | 4 | 618 | XP_011540532.1 | |
| XM_017002428.1 | 5 | 500 | XP_016857917.1 | |
| AK125085.1 | 6 | 1004 | ||
| AK301442.1 | 5 | 949 | ||
| AK313113.1 | 5 | 471 | ||
| BC024601.1 | 2 | 184 | AAH24601.1 | |
| BC107752.1 | 5 | 481 | AAI07753.1 | |
| BC109241.1 | 4 | 432 | AAI09242.1 | |
| BC109242.1 | 4 | 432 | AAI09243.1 | |
| BE870234.1 | 5 | 492 | ||
| BT019470.1 | 4 | 432 | AAV38277.1 | |
| BT019471.1 | 4 | 432 | AAV38278.1 | |
| CB267021.1 | 2 | 226 | ||
| EB387955.1 | 5 | 452 | ||
| U03271.1 | 4 | 432 | AAA87395.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv21e212 | Chr.1:19324106 - 19361938 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map