Assay Details
Target Gene Details
Entrez Gene ID: | 23339 |
Gene Name: | VPS39, HOPS complex subunit |
Gene Aliases: |
TLP, VAM6, hVam6p |
Location: |
Chr.15:42158701-42208331 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VPS39 | NM_001301138.1 | 26 | 3622 | NP_001288067.1 |
| NM_015289.3 | 25 | 3589 | NP_056104.2 | |
| XM_011521403.2 | 26 | 3661 | XP_011519705.1 | |
| XM_011521404.2 | 25 | 3664 | XP_011519706.1 | |
| XM_017022038.1 | 25 | 3777 | XP_016877527.1 | |
| XM_017022039.1 | 25 | 3588 | XP_016877528.1 | |
| AB018313.2 | 25 | 3540 | ||
| AF281052.1 | 25 | 3559 | AAQ05978.1 | |
| AK025334.1 | 10 | 1789 | ||
| AK074193.1 | 1 | 81 | ||
| AK124255.1 | 1 | 1582 | ||
| AK128403.1 | 8 | 2911 | ||
| AL117456.1 | 1 | 676 | ||
| BC015817.2 | 21 | 3149 | AAH15817.2 | |
| BX537404.1 | 25 | 3568 | CAD97646.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9245 | Chr.15:42149651 - 42161374 on Build GRCh38 | Gain |
|
| nsv952614 | Chr.15:42139503 - 42164202 on Build GRCh38 | Deletion |
|
| nsv471683 | Chr.15:42042177 - 42210784 on Build GRCh38 | Gain+Loss |
|
| nsv832985 | Chr.15:42079284 - 42272768 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map