Assay Details
Target Gene Details
Entrez Gene ID: | 83666 |
Gene Name: | poly(ADP-ribose) polymerase family member 9 |
Gene Aliases: |
ARTD9, BAL, BAL1, MGC:7868 |
Location: |
Chr.3:122527910-122564676 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PARP9 | NM_001146102.1 | 11 | 2475 | NP_001139574.1 |
| NM_001146103.1 | 11 | 2552 | NP_001139575.1 | |
| NM_001146104.1 | 11 | 2551 | NP_001139576.1 | |
| NM_001146105.1 | 11 | 2370 | NP_001139577.1 | |
| NM_031458.2 | 11 | 2657 | NP_113646.2 | |
| XM_005247820.2 | 11 | 2581 | XP_005247877.1 | |
| XM_011513218.1 | XP_011511520.1 | |||
| XM_017007304.1 | 11 | 2465 | XP_016862793.1 | |
| XM_017007305.1 | 11 | 2360 | XP_016862794.1 | |
| XM_017007306.1 | 11 | 2441 | XP_016862795.1 | |
| AF307338.1 | 11 | 2558 | AAK07558.1 | |
| AF307339.1 | 11 | 2453 | AAK07559.1 | |
| AK123412.1 | 4 | 2004 | ||
| AK130147.1 | 6 | 1006 | ||
| AK292959.1 | 11 | 2330 | ||
| AK299107.1 | 11 | 2444 | ||
| AK313494.1 | 11 | 2444 | ||
| AL713679.1 | 4 | 758 | CAD28483.1 | |
| AL832929.1 | 4 | 690 | CAH56290.1 | |
| BC039580.1 | 10 | 2328 | AAH39580.1 | |
| BX648869.1 | 9 | 1197 | CAH56133.1 |
More Information
Set Membership: |
|

Genomic Map