Assay Details
Target Gene Details
Entrez Gene ID: | 285704 |
Gene Name: | repulsive guidance molecule family member b |
Gene Aliases: |
DRAGON |
Location: |
Chr.5:98768386-98796494 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RGMB | NM_001012761.2 | 1 | 123 | NP_001012779.2 |
| XM_005271966.3 | XP_005272023.1 | |||
| XM_017009390.1 | 1 | 126 | XP_016864879.1 | |
| XM_017009391.1 | XP_016864880.1 | |||
| AK074887.1 | 1 | 125 | BAC11268.1 | |
| BC067736.1 | 1 | 123 | AAH67736.1 | |
| BI763873.1 |
More Information
Set Membership: |
|

Genomic Map