Assay Details
Target Gene Details
Entrez Gene ID: | 1581 |
Gene Name: | cytochrome P450 family 7 subfamily A member 1 |
Gene Aliases: |
CP7A, CYP7, CYPVII |
Location: |
Chr.8:58490178-58500161 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP7A1 | NM_000780.3 | 3 | 883 | NP_000771.2 |
| BC101777.1 | 3 | 883 | AAI01778.1 | |
| BC112184.1 | 3 | 883 | AAI12185.1 | |
| M93133.1 | 3 | 879 | AAA58435.1 | |
| X56088.1 | 3 | 859 | CAA39568.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv994118 | Chr.8:58489845 - 58497470 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map