Assay Details
Target Gene Details
Entrez Gene ID: | 5470 |
Gene Name: | protein phosphatase with EF-hand domain 2 |
Gene Aliases: |
PPP7CB |
Location: |
Chr.4:75859872-75902528 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPEF2 | NM_006239.2 | 17 | 2454 | NP_006230.2 |
| XM_011532039.2 | 16 | 2687 | XP_011530341.1 | |
| AF023456.1 | 17 | 2464 | AAB82796.1 | |
| BX648754.1 | 18 | 3197 |
More Information
Set Membership: |
|

Genomic Map