Assay Details
Target Gene Details
Entrez Gene ID: | 8999 |
Gene Name: | cyclin dependent kinase like 2 |
Gene Aliases: |
KKIAMRE, P56 |
Location: |
Chr.4:75576494-75630537 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDKL2 | NM_003948.3 | NP_003939.1 | ||
| XM_005263335.2 | XP_005263392.1 | |||
| XM_006714406.2 | XP_006714469.1 | |||
| XM_017008809.1 | XP_016864298.1 | |||
| XM_017008810.1 | XP_016864299.1 | |||
| XM_017008811.1 | XP_016864300.1 | |||
| AK294866.1 | ||||
| AK309127.1 | ||||
| AK312490.1 | ||||
| BC093646.1 | AAH93646.1 | |||
| BC093981.1 | AAH93981.1 | |||
| U35146.1 | AAC50918.1 |
More Information
Set Membership: |
|

Genomic Map