Assay Details
Target Gene Details
Entrez Gene ID: | 26160 |
Gene Name: | intraflagellar transport 172 |
Gene Aliases: |
BBS20, NPHP17, RP71, SLB, SRTD10, osm-1, wim |
Location: |
Chr.2:27444373-27489819 on Build GRCh38 |
Assay Gene Location: | Within Exon 43 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFT172 | NM_015662.2 | 39 | 4419 | NP_056477.1 |
| XM_005264254.1 | 39 | 4250 | XP_005264311.1 | |
| XM_006711986.3 | 40 | 4511 | XP_006712049.1 | |
| XM_006711987.1 | 39 | 4316 | XP_006712050.1 | |
| XM_011532757.2 | 39 | 4625 | XP_011531059.1 | |
| XM_011532759.2 | 25 | 2901 | XP_011531061.1 | |
| XM_011532760.2 | 22 | 2581 | XP_011531062.1 | |
| XM_017003790.1 | 40 | 4445 | XP_016859279.1 | |
| XM_017003791.1 | 38 | 4386 | XP_016859280.1 | |
| XM_017003793.1 | 35 | 3826 | XP_016859282.1 | |
| XM_017003794.1 | 33 | 3529 | XP_016859283.1 | |
| XM_017003795.1 | 37 | 4378 | XP_016859284.1 | |
| AB033005.1 | 21 | 2284 | BAA86493.1 | |
| AL110218.1 | 36 | 3975 | CAB53678.1 | |
| AL117421.1 | 7 | 665 | CAB55914.2 | |
| BC008024.1 | 7 | 617 | AAH08024.1 | |
| BC047294.1 | 7 | 716 | AAH47294.1 | |
| BC137126.1 | 39 | 4309 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833614 | Chr.2:27263559 - 27452213 on Build GRCh38 | Gain |
|
| nsv817684 | Chr.2:27434300 - 27623736 on Build GRCh38 | Gain |
|
| nsv581244 | Chr.2:27335385 - 27480628 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map