Assay Details
Target Gene Details
Entrez Gene ID: | 25852 |
Gene Name: | armadillo repeat containing 8 |
Gene Aliases: |
GID5, HSPC056, S863-2, VID28 |
Location: |
Chr.3:138187248-138298389 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 16 - Intron 16 |
Target Gene Details
Entrez Gene ID: | 347736 |
Gene Name: | NME/NM23 family member 9 |
Gene Aliases: |
NM23-H9, TXL-2, TXL2, TXNDC6 |
Location: |
Chr.3:138261437-138331785 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NME9 | NM_178130.3 | NP_835231.1 | ||
| AK094830.1 | BAC04430.1 | |||
| AY336746.1 | AAQ16116.1 | |||
| BC107054.2 | AAI07055.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428422 | Chr.3:137344347 - 138686784 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map