Assay Details
Target Gene Details
Entrez Gene ID: | 5501 |
Gene Name: | protein phosphatase 1 catalytic subunit gamma |
Gene Aliases: |
PP-1G, PP1C, PPP1G |
Location: |
Chr.12:110708369-110742978 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1CC | NM_001244974.1 | 8 | 1288 | NP_001231903.1 |
| NM_002710.3 | 7 | 2188 | NP_002701.1 | |
| XM_011538504.2 | XP_011536806.1 | |||
| XM_011538505.2 | XP_011536807.1 | |||
| AA781342.1 | 1 | 227 | ||
| AK091841.1 | 1 | 1913 | ||
| AW575661.1 | 1 | 339 | ||
| L07395.1 | 7 | 1905 | AAA19823.1 | |
| X74008.1 | 7 | 2071 | CAA52169.1 |
More Information
Set Membership: |
|

Genomic Map