Assay Details
Target Gene Details
Entrez Gene ID: | 65057 |
Gene Name: | adrenocortical dysplasia homolog |
Gene Aliases: |
PIP1, PTOP, TINT1, TPP1 |
Location: |
Chr.16:67657512-67660815 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACD | NM_001082486.1 | 2 | 786 | NP_001075955.1 |
| NM_001082487.1 | 2 | 777 | NP_001075956.1 | |
| NM_022914.2 | 2 | 777 | NP_075065.2 | |
| XM_005256115.3 | 2 | 786 | XP_005256172.1 | |
| AF070535.1 | 2 | 186 | ||
| AK023726.1 | 2 | 772 | BAB14658.1 | |
| AK125623.1 | 1 | 318 | ||
| AK301090.1 | 2 | 203 | ||
| AY502940.1 | 2 | 452 | AAS80318.1 | |
| BC016904.1 | 2 | 714 | AAH16904.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain |
|
| nsv952046 | Chr.16:67628798 - 67678697 on Build GRCh38 | Deletion |
|
| nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map