Assay Details
Target Gene Details
Entrez Gene ID: | 3163 |
Gene Name: | heme oxygenase 2 |
Gene Aliases: |
HO-2 |
Location: |
Chr.16:4474697-4510347 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HMOX2 | NM_001127204.1 | 5 | 907 | NP_001120676.1 |
| NM_001127205.1 | 4 | 811 | NP_001120677.1 | |
| NM_001127206.2 | 4 | 786 | NP_001120678.1 | |
| NM_001286267.1 | 5 | 910 | NP_001273196.1 | |
| NM_001286268.1 | 4 | 967 | NP_001273197.1 | |
| NM_001286269.1 | 5 | 888 | NP_001273198.1 | |
| NM_001286270.1 | 6 | 1187 | NP_001273199.1 | |
| NM_001286271.1 | 3 | 662 | NP_001273200.1 | |
| NM_002134.3 | 4 | 789 | NP_002125.3 | |
| XM_011522473.1 | 4 | 777 | XP_011520775.1 | |
| XM_017023196.1 | 5 | 831 | XP_016878685.1 | |
| XM_017023197.1 | 4 | 662 | XP_016878686.1 | |
| AF051306.1 | AAC05297.1 | |||
| AK093388.1 | 6 | 1148 | ||
| AK222629.1 | 4 | 681 | BAD96349.1 | |
| AK313660.1 | 4 | 741 | ||
| BC002396.2 | 4 | 705 | AAH02396.1 | |
| BE561721.1 | ||||
| BM905872.1 | 3 | 567 | ||
| BT019788.1 | 3 | 600 | AAV38591.1 | |
| CR456760.1 | 3 | 600 | CAG33041.1 | |
| CR541839.1 | 3 | 600 | CAG46638.1 | |
| D21243.1 | 3 | 600 | BAA04789.1 | |
| S34389.1 | 4 | 684 | AAB22110.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map