Assay Details
Target Gene Details
Entrez Gene ID: | 4109 |
Gene Name: | MAGE family member A10 |
Gene Aliases: |
CT1.10, MAGE10 |
Location: |
Chr.X:152133310-152138578 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEA10 | NM_001011543.2 | 5 | 415 | NP_001011543.2 |
| NM_001251828.1 | 5 | 343 | NP_001238757.1 | |
| NM_021048.4 | 4 | 324 | NP_066386.2 | |
| AK314161.1 | 4 | 324 | ||
| BC004105.2 | 4 | 299 | AAH04105.1 | |
| CB158974.1 | 5 | 383 | ||
| CB959556.1 | 5 | 426 | ||
| DA827990.1 | 5 | 305 |
Target Gene Details
Entrez Gene ID: | 100533997 |
Gene Name: | MAGEA10-MAGEA5 readthrough |
Gene Aliases: |
CT1.5 |
Location: |
Chr.X:152114049-152138578 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEA10-MAGEA5 | NM_001204811.2 | NP_001191740.1 | ||
| BX109673.1 | ||||
| FY212575.1 | ||||
| FY212810.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520971 | Chr.X:152109627 - 152136472 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map