Assay Details
Target Gene Details
Entrez Gene ID: | 6787 |
Gene Name: | NIMA related kinase 4 |
Gene Aliases: |
NRK2, STK2, pp12301 |
Location: |
Chr.3:52707936-52770949 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEK4 | XM_006713310.2 | 15 | 6176 | XP_006713373.1 |
Target Gene Details
Entrez Gene ID: | 28972 |
Gene Name: | signal peptidase complex subunit 1 |
Gene Aliases: |
HSPC033, SPC1, SPC12, YJR010C-A |
Location: |
Chr.3:52705841-52708182 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPCS1 | NM_014041.3 | NP_054760.3 | ||
| AF092138.1 | AAD40380.1 | |||
| AK027426.1 | ||||
| AX775783.1 | CAE11591.1 | |||
| AX775785.1 | CAE11592.1 | |||
| AX775787.1 | CAE11593.1 | |||
| AX775833.1 | CAE11616.1 | |||
| BC000884.1 | AAH00884.1 | |||
| BI826431.1 | 4 | 786 | ||
| BQ423345.1 | ||||
| CA309210.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508219 | Chr.3:52707177 - 52769883 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map