Assay Details
Target Gene Details
Entrez Gene ID: | 3992 |
Gene Name: | fatty acid desaturase 1 |
Gene Aliases: |
D5D, FADS6, FADSD5, LLCDL1, TU12 |
Location: |
Chr.11:61799625-61817057 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FADS1 | NM_013402.4 | 1 | 403 | NP_037534.3 |
| AF084558.1 | 1 | 182 | AAG23120.1 | |
| AF199596.1 | 1 | 104 | AAF29378.1 | |
| AF226273.1 | 1 | 104 | AAF70457.1 | |
| AK027427.1 | 1 | 113 | BAB55103.1 | |
| AK027522.1 | 1 | 203 | BAB55173.1 | |
| AK074754.1 | 1 | 342 | BAC11182.1 | |
| AK074819.1 | 1 | 337 | BAC11229.1 | |
| AK222906.1 | 1 | 147 | BAD96626.1 | |
| AK314199.1 | 1 | 187 | ||
| AL512760.1 | 1 | 184 | CAC21679.1 | |
| BC007846.2 | 1 | 126 | AAH07846.1 | |
| BP253788.1 | 1 | 429 |
Target Gene Details
Entrez Gene ID: | 9415 |
Gene Name: | fatty acid desaturase 2 |
Gene Aliases: |
D6D, DES6, FADSD6, LLCDL2, SLL0262, TU13 |
Location: |
Chr.11:61816203-61867354 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FADS2 | NM_001281501.1 | NP_001268430.1 | ||
| NM_001281502.1 | NP_001268431.1 | |||
| AF108658.1 | AAG43192.1 | |||
| AK074991.1 | ||||
| AK290291.1 | ||||
| AK299762.1 | ||||
| HY090295.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528697 | Chr.11:61690385 - 61919561 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map